siRNA Details
Gene Symbol: 9830147E19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242388.1 | NP_001229317.1 | 4 | 4154 | ||
GenBank mRNA | AK162659.1 | - | 3 | 4010 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 9830147E19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242388.1 | NP_001229317.1 | 4 | 4154 | ||
GenBank mRNA | AK162659.1 | - | 3 | 4010 |