siRNA Details
| Gene Symbol: 1810043H04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110242.1 | NP_001103712.1 | 3 | 417 | ||
| GenBank mRNA | AK007756.1 | - | 3 | 415 | ||
| AK010111.1 | - | 3 | 415 | |||
| BC061009.1 | - | 1 | 127 | |||
| BC147711.1 | - | 3 | 397 | 9087470 | ||
| BC147720.1 | - | 3 | 397 | 9087479 | ||

1810043H04Rik
Genomic Map