siRNA Details
| Gene Symbol: Sult2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111296.2 | NP_001104766.1 | 2 | 309 | ||
| GenBank mRNA | BC145167.1 | - | 2 | 281 | ||
| L02335.1 | - | 2 | 282 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sult2a1| Gene Symbol: Sult2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111296.2 | NP_001104766.1 | 2 | 309 | ||
| GenBank mRNA | BC145167.1 | - | 2 | 281 | ||
| L02335.1 | - | 2 | 282 | |||