siRNA Details
| Gene Symbol: Tmem212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164437.1 | NP_001157909.1 | 4 | 648 | ||
| GenBank mRNA | AK053134.1 | - | 4 | 648 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem212| Gene Symbol: Tmem212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164437.1 | NP_001157909.1 | 4 | 648 | ||
| GenBank mRNA | AK053134.1 | - | 4 | 648 | ||