siRNA Details
Gene Symbol: Unc13c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081153.1 | NP_001074622.1 | 2 | 2983 | ||
GenBank mRNA | BC030416.1 | - | 2 | 975 | ||
BC048861.1 | - | 2 | 964 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Unc13c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081153.1 | NP_001074622.1 | 2 | 2983 | ||
GenBank mRNA | BC030416.1 | - | 2 | 975 | ||
BC048861.1 | - | 2 | 964 |