siRNA Details
| Gene Symbol: Ccdc62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134767.1 | NP_001128239.1 | 13 | 2891 | ||
| GenBank mRNA | AK135815.1 | - | 1 | 77 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc62| Gene Symbol: Ccdc62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134767.1 | NP_001128239.1 | 13 | 2891 | ||
| GenBank mRNA | AK135815.1 | - | 1 | 77 | ||