siRNA Details
| Gene Symbol: Gan | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081151.1 | NP_001074620.1 | 2, 3 | 286 | ||
| GenBank mRNA | AK039455.1 | - | 2, 3 | 285 | ||
| BC112432.1 | - | 2, 3 | 285 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gan| Gene Symbol: Gan | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081151.1 | NP_001074620.1 | 2, 3 | 286 | ||
| GenBank mRNA | AK039455.1 | - | 2, 3 | 285 | ||
| BC112432.1 | - | 2, 3 | 285 | |||