siRNA Details
Gene Symbol: Csta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033239.3 | NP_001028411.1 | 3 | 2664 | ||
GenBank mRNA | AK137702.1 | - | 3 | 2665 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Csta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033239.3 | NP_001028411.1 | 3 | 2664 | ||
GenBank mRNA | AK137702.1 | - | 3 | 2665 |