siRNA Details
| Gene Symbol: Wfdc6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033240.4 | NP_001028412.1 | 3 | 184 | ||
| NM_001177848.1 | NP_001171319.1 | 2 | 193 | |||
| GenBank mRNA | AK136589.1 | - | 2 | 194 | ||
| BC147597.1 | - | 2 | 167 | 9056587 | ||
| BC147615.1 | - | 2 | 167 | 9056605 | ||
| BM933122.1 | - | 3 | 186 | |||

Wfdc6a
Genomic Map