siRNA Details
| Gene Symbol: Vmn1r183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203489.1 | NP_987074.1 | 1 | 250 | ||
| GenBank mRNA | BC130237.1 | - | 1 | 254 | 40142092 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r183| Gene Symbol: Vmn1r183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203489.1 | NP_987074.1 | 1 | 250 | ||
| GenBank mRNA | BC130237.1 | - | 1 | 254 | 40142092 | |