siRNA Details
| Gene Symbol: Zfp719 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172482.1 | NP_766070.1 | 4 | 856 | ||
| GenBank mRNA | AK033195.1 | - | 1 | 235 | ||
| AK044684.1 | - | 4 | 833 | |||
| AK083143.1 | - | 4 | 797 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp719| Gene Symbol: Zfp719 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172482.1 | NP_766070.1 | 4 | 856 | ||
| GenBank mRNA | AK033195.1 | - | 1 | 235 | ||
| AK044684.1 | - | 4 | 833 | |||
| AK083143.1 | - | 4 | 797 | |||