siRNA Details
| Gene Symbol: Shank2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081370.2 | NP_001074839.2 | 11 | 5184 | ||
| NM_001113373.2 | NP_001106844.2 | 16 | 5630 | |||
| GenBank mRNA | AK042463.1 | - | 1 | 296 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Shank2| Gene Symbol: Shank2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081370.2 | NP_001074839.2 | 11 | 5184 | ||
| NM_001113373.2 | NP_001106844.2 | 16 | 5630 | |||
| GenBank mRNA | AK042463.1 | - | 1 | 296 | ||