siRNA Details
| Gene Symbol: Uhmk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017293.1 | NP_058989.1 | 1, 2 | 429 | ||
| GenBank mRNA | BC098669.1 | - | 1, 2 | 424 | 7096907 | |
| U70372.2 | - | 1, 2 | 429 | |||
| X98374.1 | - | 1, 2 | 306 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uhmk1| Gene Symbol: Uhmk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017293.1 | NP_058989.1 | 1, 2 | 429 | ||
| GenBank mRNA | BC098669.1 | - | 1, 2 | 424 | 7096907 | |
| U70372.2 | - | 1, 2 | 429 | |||
| X98374.1 | - | 1, 2 | 306 | |||