siRNA Details
| Gene Symbol: Tmem151b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013749.2 | NP_001013771.1 | 3 | 2294 | ||
| GenBank mRNA | BC079879.1 | - | 3 | 2294 | 6852149 | |
| BC083182.1 | - | 3 | 1915 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem151b| Gene Symbol: Tmem151b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013749.2 | NP_001013771.1 | 3 | 2294 | ||
| GenBank mRNA | BC079879.1 | - | 3 | 2294 | 6852149 | |
| BC083182.1 | - | 3 | 1915 | |||