siRNA Details
| Gene Symbol: Tmem151b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013749.2 | NP_001013771.1 | 3 | 3084 | ||
| GenBank mRNA | AK137932.1 | - | 1 | 695 | ||
| BC079879.1 | - | 3 | 3085 | 6852149 | ||
| BC083182.1 | - | 3 | 2704 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem151b| Gene Symbol: Tmem151b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013749.2 | NP_001013771.1 | 3 | 3084 | ||
| GenBank mRNA | AK137932.1 | - | 1 | 695 | ||
| BC079879.1 | - | 3 | 3085 | 6852149 | ||
| BC083182.1 | - | 3 | 2704 | |||