siRNA Details
| Gene Symbol: Coq10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081040.1 | NP_001074509.1 | 5 | 2272 | ||
| GenBank mRNA | AK185466.1 | - | 1 | 367 | ||
| AK205889.1 | - | 1 | 364 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Coq10a| Gene Symbol: Coq10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081040.1 | NP_001074509.1 | 5 | 2272 | ||
| GenBank mRNA | AK185466.1 | - | 1 | 367 | ||
| AK205889.1 | - | 1 | 364 | |||