siRNA Details
Gene Symbol: 6720489N17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173381.1 | NP_775557.1 | 3 | 2356 | ||
GenBank mRNA | AK033958.1 | - | 3 | 2340 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 6720489N17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173381.1 | NP_775557.1 | 3 | 2356 | ||
GenBank mRNA | AK033958.1 | - | 3 | 2340 |