siRNA Details
| Gene Symbol: 6720489N17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173381.1 | NP_775557.1 | 3 | 2184 | ||
| GenBank mRNA | AK033958.1 | - | 3 | 2168 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

6720489N17Rik| Gene Symbol: 6720489N17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173381.1 | NP_775557.1 | 3 | 2184 | ||
| GenBank mRNA | AK033958.1 | - | 3 | 2168 | ||