siRNA Details
Gene Symbol: Ccdc114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033243.2 | NP_001028415.1 | 1 | 89 | ||
GenBank mRNA | AK135860.1 | - | 1 | 90 | ||
BC172013.1 | - | 1 | 85 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ccdc114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033243.2 | NP_001028415.1 | 1 | 89 | ||
GenBank mRNA | AK135860.1 | - | 1 | 90 | ||
BC172013.1 | - | 1 | 85 |