siRNA Details
Gene Symbol: Ptchd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093750.1 | NP_001087219.1 | 3 | 5388 | ||
GenBank mRNA | AK135103.1 | - | 1 | 1633 | ||
AK141827.1 | - | 1 | 1629 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ptchd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093750.1 | NP_001087219.1 | 3 | 5388 | ||
GenBank mRNA | AK135103.1 | - | 1 | 1633 | ||
AK141827.1 | - | 1 | 1629 |