siRNA Details
| Gene Symbol: Asxl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167777.1 | NP_001161249.1 | 14 | 9470 | ||
| GenBank mRNA | AK052399.1 | - | 1 | 658 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Asxl3| Gene Symbol: Asxl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167777.1 | NP_001161249.1 | 14 | 9470 | ||
| GenBank mRNA | AK052399.1 | - | 1 | 658 | ||