siRNA Details
| Gene Symbol: Zfp748 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035231.3 | NP_001030308.1 | 4 | 4008 | ||
| GenBank mRNA | AK132196.1 | - | 4 | 3995 | ||
| AK220562.1 | - | 4 | 3988 | |||
| BC055310.1 | - | 4 | 3994 | 30048242 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp748| Gene Symbol: Zfp748 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035231.3 | NP_001030308.1 | 4 | 4008 | ||
| GenBank mRNA | AK132196.1 | - | 4 | 3995 | ||
| AK220562.1 | - | 4 | 3988 | |||
| BC055310.1 | - | 4 | 3994 | 30048242 | ||