siRNA Details
| Gene Symbol: 9330159F19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162537.2 | NP_001156009.1 | 5 | 1285 | ||
| GenBank mRNA | AK034141.1 | - | 6 | 1306 | ||
| BC138279.1 | - | 5 | 1223 | |||
| BC138282.1 | - | 5 | 1237 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9330159F19Rik| Gene Symbol: 9330159F19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162537.2 | NP_001156009.1 | 5 | 1285 | ||
| GenBank mRNA | AK034141.1 | - | 6 | 1306 | ||
| BC138279.1 | - | 5 | 1223 | |||
| BC138282.1 | - | 5 | 1237 | |||