siRNA Details
| Gene Symbol: Gm17296 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159907.1 | NP_001153379.1 | 14 | 2216 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm17296| Gene Symbol: Gm17296 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159907.1 | NP_001153379.1 | 14 | 2216 | ||