siRNA Details
| Gene Symbol: Rsph4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162957.1 | NP_001156429.1 | 2 | 866 | ||
| GenBank mRNA | AK038980.1 | - | 2 | 493 | ||
| BC138000.1 | - | 2 | 866 | 8861019 | ||
| BC138001.1 | - | 2 | 866 | 8861020 | ||

Rsph4a
Genomic Map