siRNA Details
| Gene Symbol: Armc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034858.3 | NP_001030030.2 | 3 | 468 | ||
| GenBank mRNA | BM942469.1 | - | 3 | 451 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Armc2| Gene Symbol: Armc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034858.3 | NP_001030030.2 | 3 | 468 | ||
| GenBank mRNA | BM942469.1 | - | 3 | 451 | ||