siRNA Details
| Gene Symbol: Gm732 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033252.2 | NP_001028424.1 | 1 | 101 | ||
| GenBank mRNA | AK132985.1 | - | 1 | 103 | ||
| BC147603.1 | - | 1 | 40 | 9056593 | ||
| BC147609.1 | - | 1 | 40 | 9056599 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm732| Gene Symbol: Gm732 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033252.2 | NP_001028424.1 | 1 | 101 | ||
| GenBank mRNA | AK132985.1 | - | 1 | 103 | ||
| BC147603.1 | - | 1 | 40 | 9056593 | ||
| BC147609.1 | - | 1 | 40 | 9056599 | ||