siRNA Details
| Gene Symbol: Plekhg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033253.3 | NP_001028425.1 | 16 | 5495 | ||
| NM_001159942.1 | NP_001153414.1 | 17 | 5661 | |||
| GenBank mRNA | AK076288.1 | - | 1 | 1134 | ||
| AK122468.1 | - | 13 | 4819 | |||
| BC076628.1 | - | 16 | 3680 | |||

Plekhg1
Genomic Map