siRNA Details
| Gene Symbol: Hhipl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044380.1 | NP_001037845.1 | 8, 9 | 1981 | ||
| GenBank mRNA | AK129450.1 | - | 1376 | |||
| AK147996.1 | - | 1648 | ||||
| AK166269.1 | - | 1877 | ||||
| BC057108.1 | - | 974 | ||||
| BC113163.1 | - | 8, 9 | 1981 | 40060054 | ||
| BC113773.1 | - | 8, 9 | 1977 | 40060051 | ||

Hhipl1
Genomic Map