siRNA Details
| Gene Symbol: Hhipl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044380.1 | NP_001037845.1 | 2 | 597 | ||
| GenBank mRNA | AK147996.1 | - | 264 | |||
| AK166269.1 | - | 493 | ||||
| BC113163.1 | - | 2 | 597 | 40060054 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hhipl1| Gene Symbol: Hhipl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044380.1 | NP_001037845.1 | 2 | 597 | ||
| GenBank mRNA | AK147996.1 | - | 264 | |||
| AK166269.1 | - | 493 | ||||
| BC113163.1 | - | 2 | 597 | 40060054 | ||