siRNA Details
| Gene Symbol: BC003965 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183150.2 | NP_898973.2 | 2 | 1969 | ||
| GenBank mRNA | BC094389.1 | - | 1 | 2035 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC003965| Gene Symbol: BC003965 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183150.2 | NP_898973.2 | 2 | 1969 | ||
| GenBank mRNA | BC094389.1 | - | 1 | 2035 | ||