siRNA Details
| Gene Symbol: Gm136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033255.2 | NP_001028427.1 | 4 | 771 | ||
| GenBank mRNA | AK133458.1 | - | 4 | 772 | ||
| BC147519.1 | - | 4 | 763 | 9056511 | ||
| BC147522.1 | - | 4 | 763 | 9056514 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm136| Gene Symbol: Gm136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033255.2 | NP_001028427.1 | 4 | 771 | ||
| GenBank mRNA | AK133458.1 | - | 4 | 772 | ||
| BC147519.1 | - | 4 | 763 | 9056511 | ||
| BC147522.1 | - | 4 | 763 | 9056514 | ||