siRNA Details
| Gene Symbol: Tdrd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134741.1 | NP_001128213.1 | 14 | 2230 | ||
| NM_001277730.1 | NP_001264659.1 | 14 | 1936 | |||
| GenBank mRNA | AK132674.1 | - | 14 | 2215 | ||
| AK133026.1 | - | 12 | 1602 | |||
| AK138828.1 | - | 3 | 3356 | |||
| AY522557.1 | - | 8 | 1090 | |||
| BC099972.1 | - | 14 | 1936 | 30080156 | ||

Tdrd5
Genomic Map