siRNA Details
| Gene Symbol: Chadl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164320.1 | NP_001157792.1 | 3 | 257 | ||
| GenBank mRNA | AK190820.1 | - | 2 | 65 | ||
| AY100452.1 | - | 3 | 313 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chadl| Gene Symbol: Chadl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164320.1 | NP_001157792.1 | 3 | 257 | ||
| GenBank mRNA | AK190820.1 | - | 2 | 65 | ||
| AY100452.1 | - | 3 | 313 | |||