siRNA Details
| Gene Symbol: Gm4791 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243258.1 | NP_001230187.1 | 3 | 788 | ||
| GenBank mRNA | AK086549.1 | - | 3 | 789 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm4791| Gene Symbol: Gm4791 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243258.1 | NP_001230187.1 | 3 | 788 | ||
| GenBank mRNA | AK086549.1 | - | 3 | 789 | ||