siRNA Details
| Gene Symbol: Rfpl4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177783.1 | NP_001171254.1 | 2 | 248 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY726306.1 | - | 248 | Chr. - - [-] on Build | ||

Rfpl4b
Genomic Map