siRNA Details
| Gene Symbol: Stox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033260.1 | NP_001028432.1 | 4 | 2974 | ||
| GenBank mRNA | AK028920.1 | - | 4 | 2741 | ||
| AK133671.1 | - | 2 | 1562 | |||
| BC137978.1 | - | 4 | 2974 | 8860997 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stox1| Gene Symbol: Stox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033260.1 | NP_001028432.1 | 4 | 2974 | ||
| GenBank mRNA | AK028920.1 | - | 4 | 2741 | ||
| AK133671.1 | - | 2 | 1562 | |||
| BC137978.1 | - | 4 | 2974 | 8860997 | ||