siRNA Details
| Gene Symbol: Trappc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081055.1 | NP_001074524.1 | 23 | 4844 | ||
| GenBank mRNA | AK154290.1 | - | 23 | 4781 | ||
| BC044902.1 | - | 15 | 3500 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trappc10| Gene Symbol: Trappc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081055.1 | NP_001074524.1 | 23 | 4844 | ||
| GenBank mRNA | AK154290.1 | - | 23 | 4781 | ||
| BC044902.1 | - | 15 | 3500 | |||