siRNA Details
| Gene Symbol: Agap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033263.5 | NP_001028435.1 | 19 | 5040 | ||
| NM_001301014.1 | NP_001287943.1 | 18 | 4980 | |||
| GenBank mRNA | AK049954.1 | - | 1 | 62 | ||
| AK147446.1 | - | 19 | 5041 | |||
| AK220325.1 | - | 18 | 3854 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_023026.

Agap2
Genomic Map