siRNA Details
| Gene Symbol: Clint1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045520.3 | NP_001038985.2 | 7 | 957 | ||
| GenBank mRNA | AB093212.1 | - | 6 | 2044 | ||
| AK143544.1 | - | 7 | 958 | |||
| AK168991.1 | - | 7 | 923 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clint1| Gene Symbol: Clint1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045520.3 | NP_001038985.2 | 7 | 957 | ||
| GenBank mRNA | AB093212.1 | - | 6 | 2044 | ||
| AK143544.1 | - | 7 | 958 | |||
| AK168991.1 | - | 7 | 923 | |||