siRNA Details
| Gene Symbol: Trim58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039047.1 | NP_001034136.1 | 6 | 1234 | ||
| GenBank mRNA | BC147805.1 | - | 6 | 1253 | 9087522 | |
| BC147806.1 | - | 6 | 1253 | 9087523 | ||
| BC150936.1 | - | 6 | 1308 | 9087847 | ||

Trim58
Genomic Map