siRNA Details
| Gene Symbol: Heatr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145432.3 | NP_663407.2 | 6 | 815 | ||
| GenBank mRNA | AK012216.1 | - | 6 | 759 | ||
| AK076219.1 | - | 6 | 943 | |||
| AK132494.1 | - | 6 | 750 | |||
| AK149059.1 | - | 6 | 609 | |||
| AK158323.1 | - | 6 | 945 | |||
| AK169901.1 | - | 5 | 2242 | |||
| BC065092.1 | - | 6 | 741 | 6855410 | ||

Heatr6
Genomic Map