siRNA Details
| Gene Symbol: Heatr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145432.3 | NP_663407.2 | 20 | 4717 | ||
| GenBank mRNA | AK136845.1 | - | 4 | 2069 | ||
| AK138219.1 | - | 4 | 1975 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Heatr6| Gene Symbol: Heatr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145432.3 | NP_663407.2 | 20 | 4717 | ||
| GenBank mRNA | AK136845.1 | - | 4 | 2069 | ||
| AK138219.1 | - | 4 | 1975 | |||