siRNA Details
Gene Symbol: Heatr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145432.3 | NP_663407.2 | 20 | 4567 | ||
GenBank mRNA | AK136845.1 | - | 4 | 1919 | ||
AK138219.1 | - | 4 | 1825 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Heatr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145432.3 | NP_663407.2 | 20 | 4567 | ||
GenBank mRNA | AK136845.1 | - | 4 | 1919 | ||
AK138219.1 | - | 4 | 1825 |