siRNA Details
| Gene Symbol: Gm525 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033266.2 | NP_001028438.1 | 3 | 307 | ||
| GenBank mRNA | AA237157.1 | - | 3 | 203 | ||
| AK040024.1 | - | 3 | 225 | |||
| AW823710.1 | - | 3 | 307 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm525| Gene Symbol: Gm525 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033266.2 | NP_001028438.1 | 3 | 307 | ||
| GenBank mRNA | AA237157.1 | - | 3 | 203 | ||
| AK040024.1 | - | 3 | 225 | |||
| AW823710.1 | - | 3 | 307 | |||