siRNA Details
| Gene Symbol: Gm525 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033266.2 | NP_001028438.1 | 4 | 563 | ||
| GenBank mRNA | AA237157.1 | - | 4 | 459 | ||
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 Gm525
Gm525| Gene Symbol: Gm525 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033266.2 | NP_001028438.1 | 4 | 563 | ||
| GenBank mRNA | AA237157.1 | - | 4 | 459 | ||
 Back To Top
 Back To Top