siRNA Details
| Gene Symbol: Cisd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085500.2 | NP_001078969.2 | 3 | 580 | ||
| GenBank mRNA | BC056217.1 | - | 3 | 575 | ||
| BC147739.1 | - | 4 | 675 | 9056688 | ||
| BC147742.1 | - | 4 | 675 | 9056691 | ||
| BC157903.1 | - | 4 | 618 | |||

Cisd3
Genomic Map