siRNA Details
| Gene Symbol: Atxn7l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098836.1 | NP_001092306.1 | 3 | 407 | ||
| NM_001098837.1 | NP_001092307.1 | 3 | 407 | |||
| GenBank mRNA | AK090368.1 | - | 3 | 409 | ||
| BC130269.1 | - | 2 | 159 | |||

Atxn7l3
Genomic Map