siRNA Details
| Gene Symbol: Slc16a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080934.1 | NP_001074403.1 | 4 | 1470 | ||
| GenBank mRNA | AK037416.1 | - | 3 | 1582 | ||
| AK079757.1 | - | 2 | 515 | |||
| BC052932.1 | - | 3 | 503 | |||
| BC053982.1 | - | 3 | 503 | |||
| BC172170.1 | - | 4 | 1485 | |||

Slc16a5
Genomic Map