siRNA Details
| Gene Symbol: Rnf157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027258.1 | NP_081534.1 | 19 | 2841 | ||
| GenBank mRNA | AK122571.1 | - | 9 | 1577 | ||
| AK160809.1 | - | 1 | 2294 | |||
| BC053070.1 | - | 19 | 2620 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf157| Gene Symbol: Rnf157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027258.1 | NP_081534.1 | 19 | 2841 | ||
| GenBank mRNA | AK122571.1 | - | 9 | 1577 | ||
| AK160809.1 | - | 1 | 2294 | |||
| BC053070.1 | - | 19 | 2620 | |||